missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Acetylcholine receptor subunit gamma Protein
A DNA sequence encoding the Homo sapiens (Human) Acetylcholine receptor subunit gamma, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9069-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005190.4 | |
Acetylcholine receptor subunit gamma Protein | |
Research Use Only | |
CHRNG | |
Recombinant Protein | |
Yeast | |
RNQEERLLAD LMQNYDPNLR PAERDSDVVN VSLKLTLTNL ISLNEREEAL TTNVWIEMQW CDYRLRWDPR DYEGLWVLRV PSTMVWRPDI VLENNVDGVF EVALYCNVLV SPDGCIYWLP PAIFRSACSI SVTYFPFDWQ NCSLIFQSQT YSTNEIDLQL SQEDGQTIEW IFIDPEAFTE NGEWAIQHRP AKMLLDPAAP AQEAGHQKVV FYLLIQRK | |
Greater than 90% as determined by SDS-PAGE. |
1146 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |