missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ABHD5 Protein
A DNA sequence encoding the Homo sapiens (Human) ABHD5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7831-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057090.2 | |
ABHD5 Protein | |
Research Use Only | |
ABHD5 | |
Recombinant Protein | |
E. coli | |
AAEEEEVDSA DTGERSGWLT GWLPTWCPTS ISHLKEAEEK MLKCVPCTYK KEPVRISNGN KIWTLKFSHN ISNKTPLVLL HGFGGGLGLW ALNFGDLCTN RPVYAFDLLG FGRSSRPRFD SDAEEVENQF VESIEEWRCA LGLDKMILLG HNLGGFLAAA YSLKYPSRVN HLILVEPWGF PERPDLADQD RPIPVWIRAL GAALTPFNPL AGLRIAGPFG LSLVQRLRPD FKRKYSSMFE DDTVTEYIYH CNVQTPSGET AFKNMTIPYG WAKRPMLQRI GKMHPDIPVS VIFGARSCID GNSGTSIQSL RPHSYVKTIA ILGAGHYVYA DQPEEFNQKV KEICDTVD | |
Greater than 90% as determined by SDS-PAGE. |
51099 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |