missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L35a Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L35a, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6621-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000987.2 | |
60S ribosomal protein L35a Protein | |
Research Use Only | |
RPL35A | |
Recombinant Protein | |
E. coli | |
MSGRLWSKAI FAGYKRGLRN QREHTALLKI EGVYARDETE FYLGKRCAYV YKAKNNTVTP GGKPNKTRVI WGKVTRAHGN SGMVRAKFRS NLPAKAIGHR IRVMLYPSRI | |
Greater than 90% as determined by SDS-PAGE. |
6165 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |