missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 39S ribosomal protein L11, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L11, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8279-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057134.1 | |
39S ribosomal protein L11, mitochondrial Protein | |
Research Use Only | |
MRPL11 | |
Recombinant Protein | |
E. coli | |
MSKLGRAARG LRKPEVGGVI RAIVRAGLAM PGPPLGPVLG QRGVSINQFC KEFNERTKDI KEGIPLPTKI LVKPDRTFEI KIGQPTVSYF LKAAAGIEKG ARQTGKEVAG LVTLKHVYEI ARIKAQDEAF ALQDVPLSSV VRSIIGSARS LGIRVVKDLS SEELAAFQKE RAIFLAAQKE ADLAAQEEAA K | |
Greater than 90% as determined by SDS-PAGE. |
65003 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |