Learn More
enQuireBio™ Recombinant H. pylori Vacuolating cytotoxin autotransporter Protein
A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) Vacuolating cytotoxin autotransporter, was expressed in the hosts and tags indicated. option, abo
Brand: enQuireBio™ QP8796-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_207680.1 | |
Vacuolating cytotoxin autotransporter Protein | |
Research Use Only | |
vacA | |
Recombinant Protein | |
E. coli | |
TTVIIPAIVG GIATGAAVGT VSGLLGWGLK QAEEANKTPD KPDKVWRIQA GKGFNEFPNK EYDLYRSLLS SKIDGGWDWG NAATHYWVKG GQWNKLEVDM KDAVGTYNLS GLRNFTGGDL DVNMQKATLR LGQFNGNSFT SYKDSADRTT RVDFNAKNIL IDNFLEINNR VGSGAGRKAS STVLTLQASE GITSSKNAEI SLYDGATLN | |
Greater than 90% as determined by SDS-PAGE. |
899415 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
H. pylori | |
His | |
Tris-based buffer, 50% glycerol |