missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant H. pylori Vacuolating cytotoxin autotransporter Protein
A DNA sequence encoding the Helicobacter pylori (Campylobacter pylori) Vacuolating cytotoxin autotransporter, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7651-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
Vacuolating cytotoxin autotransporter Protein | |
Research Use Only | |
vacA | |
Recombinant Protein | |
E. coli | |
KYEKPTNVWA NAIGGTSLNN GSNASLYGTS AGVDAYLNGE VEAIVGGFGS YGYSSFSNQA NSLNSGANNT NFGVYSRIFA NQHEFDFEAQ GALGSDQSSL NFKSALLQDL NQSYHYLAYS ATTRASYGYD FAFFRNALVL KPSVGVSYNH LGSTNFKSNS NQVALSNGSS SQHLFNANAN VEARYYYGDT SYFYMNAGVL QEFARFGSNN AVSLNTFKVN ATRNPLNTHA RVMMGGELQL AKEVFLNLGV VYLHNLISNA SHFASNLGMR YSF | |
Greater than 80% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
H. pylori | |
His-SUMO | |
Tris-based buffer, 50% glycerol |