missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant H. pylori Glutamate--tRNA ligase 1 Protein

A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) Glutamate--tRNA ligase 1, was expressed in the hosts and tags indicated. option, abo

Brand:  enQuireBio™ QP6981-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036920

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_207274.1
Glutamate--tRNA ligase 1 Protein
Research Use Only
gltX1
Recombinant Protein
E. coli
MSLIVTRFAP SPTGYLHIGG LRTAIFNYLF ARANQGKFFL RIEDTDLSRN SIEAANAIIE AFKWVGLEYD GEILYQSKRF EIYKEYIQKL LDEDKAYYCY MSKEELDALR EEQKARKETP RYDNRYRDFK GTPPKGIEPV VRIKVPQNEV IGFNDGVKGE VKVNTNELDD FIIARSDGTP TYNFVVTIDD ALMGITDVIR GDDHLSNTPK QIVLYKALNF KIPNFFHVPM ILNEEGQKLS KRHGATNVMD YQEMGYLKEA LVNFLARLGW SYQDKEVFSM QELLELFDPK DLNSSPSCFS WHKLNWLNAH YLKNQSVQEL LKLLKPFSFS DLSHLNPTQL DRLLDALKER SQTLKELALK IDEVLIAPVE YEEKVFKKLN QALVMPLLEK FKLELNKANF NDESALENAM RQIIEEEKIK AGSFMQPLRL ALLGKGGGIG LKEALFILGK TESVKRIEDF LKN
Greater than 90% as determined by SDS-PAGE.
898901
10 μg
Not determined.
Full Length
H. pylori
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers