missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant H. pylori cagA Protein
A DNA sequence encoding the Helicobacter pylori (Campylobacter pylori) cagA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6943-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
cagA Protein | |
Research Use Only | |
cagA | |
Recombinant Protein | |
E. coli | |
KVNAKIDRLN QIASGLGVVG QAAGFPLKRH DKVDDLSKVG LSRNQELAQK IDNLNQAVSE AKAGFFGNLE QTIDKLKDST KHNPMNLWVE SAKKVPASLS AKLDNYATNS HIRINSNIKN GAINEKATGM LTQKNPEWLK LVNDKIVAHN VGSVPLSEYD KIGFNQKNMK DYSDSFKFST KLNNAVKDTN SGFTQFLTNA FSTASYYCLA RENAEHGIKN VNTKGGFQKS | |
Greater than 80% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
H. pylori | |
His-SUMO | |
Tris-based buffer, 50% glycerol |