missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Latent membrane protein 1
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Latent membrane protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7054-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
YP_401631.1 | |
Latent membrane protein 1 | |
Research Use Only | |
LMP2 | |
Recombinant Protein | |
Yeast | |
MGSLEMVPMG AGPPSPGGDP DGYDGGNNSQ YPSASGSSGN TPTPPNDEER ESNEEPPPPY EDPYWGNGDR HSDYQPLGTQ DQSLYLGLQH DGNDGLPPPP YSPRDDSSQH IYEEAGRGSM NPVCLPVIVA PYLFWLAAIA ASCFTAS | |
Greater than 80% as determined by SDS-PAGE. |
17494212 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Epstein-Barr | |
His | |
Tris-based buffer, 50% glycerol |