missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Transketolase 1 Protein

A DNA sequence encoding the Escherichia coli (strain K12) Transketolase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7207-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047710

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

WP_000098614.1
Transketolase 1 Protein
Research Use Only
tktA
Recombinant Protein
E. coli
MSSRKELANA IRALSMDAVQ KAKSGHPGAP MGMADIAEVL WRDFLKHNPQ NPSWADRDRF VLSNGHGSML IYSLLHLTGY DLPMEELKNF RQLHSKTPGH PEVGYTAGVE TTTGPLGQGI ANAVGMAIAE KTLAAQFNRP GHDIVDHYTY AFMGDGCMME GISHEVCSLA GTLKLGKLIA FYDDNGISID GHVEGWFTDD TAMRFEAYGW HVIRDIDGHD AASIKRAVEE ARAVTDKPSL LMCKTIIGFG SPNKAGTHDS HGAPLGDAEI ALTREQLGWK YAPFEIPSEI YAQWDAKEAG QAKESAWNEK FAAYAKAYPQ EAAEFTRRMK GEMPSDFDAK AKEFIAKLQA NPAKIASRKA SQNAIEAFGP LLPEFLGGSA DLAPSNLTLW SGSKAINEDA AGNYIHYGVR EFGMTAIANG ISLHGGFLPY TSTFLMFVEY ARNAVRMAAL MKQRQVMVYT HDSIGLGEDG PTHQPVEQVA SLRVTPNMST WRPCDQVESA VAWKYGVERQ DGPTALILSR QNLAQQERTE EQLANIARGG YVLKDCAGQP ELIFIATGSE VELAVAAYEK LTAEGVKARV VSMPSTDAFD KQDAAYRESV LPKAVTARVA VEAGIADYWY KYVGLNGAIV GMTTFGESAP AELLFEEFGF TVDNVVAKAK ELL
Greater than 90% as determined by SDS-PAGE.
947420
200 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers