missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Transcription termination factor Rho Protein
A DNA sequence encoding the Escherichia coli (strain K12) Transcription termination factor Rho, was expressed in the hosts and tags indicated.
402.00€ - 2645.00€
Specifications
Accession Number | NP_418230.1 |
---|---|
Gene ID (Entrez) | 948297 |
Name | Transcription termination factor Rho Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006911
|
enQuireBio™
QP8868-EC-10UG |
10 μg |
402.00€
10µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16026911
|
enQuireBio™
QP8868-EC-200UG |
200 μg |
1600.00€
200µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16096901
|
enQuireBio™
QP8868-EC-100UG |
100 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16046911
|
enQuireBio™
QP8868-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16036911
|
enQuireBio™
QP8868-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16016911
|
enQuireBio™
QP8868-EC-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_418230.1 | |
Transcription termination factor Rho Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
948297 | |
Research Use Only | |
rho | |
Recombinant Protein | |
E. coli | |
MNLTELKNTP VSELITLGEN MGLENLARMR KQDIIFAILK QHAKSGEDIF GDGVLEILQD GFGFLRSADS SYLAGPDDIY VSPSQIRRFN LRTGDTISGK IRPPKEGERY FALLKVNEVN FDKPENARNK ILFENLTPLH ANSRLRMERG NGSTEDLTAR VLDLASPIGR GQRGLIVAPP KAGKTMLLQN IAQSIAYNHP DCVLMVLLID ERPEEVTEMQ RLVKGEVVAS TFDEPASRHV QVAEMVIEKA KRLVEHKKDV IILLDSITRL ARAYNTVVPA SGKVLTGGVD ANALHRPKRF FGAARNVEEG GSLTIIATAL IDTGSKMDEV IYEEFKGTGN MELHLSRKIA EKRVFPAIDY NRSGTRKEEL LTTQEELQKM WILRKIIHPM GEIDAMEFLI NKLAMTKTND DFFEMMKRS | |
Greater than 90% as determined by SDS-PAGE. |