missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli plsC Protein
A DNA sequence encoding the Escherichia coli (strain K12) plsC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7206-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417490.1 | |
plsC Protein | |
Research Use Only | |
plsC | |
Recombinant Protein | |
Yeast | |
MLYIFRLIIT VIYSILVCVF GSIYCLFSPR NPKHVATFGH MFGRLAPLFG LKVECRKPTD AESYGNAIYI ANHQNNYDMV TASNIVQPPT VTVGKKSLLW IPFFGQLYWL TGNLLIDRNN RTKAHGTIAE VVNHFKKRRI SIWMFPEGTR SRGRGLLPFK TGAFHAAIAA GVPIIPVCVS TTSNKINLNR LHNGLVIVEM LPPIDVSQYG KDQVRELAAH CRSIMEQKIA ELDKEVAERE AAGKV | |
Greater than 90% as determined by SDS-PAGE. |
947496 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |