missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli plsC Protein

A DNA sequence encoding the Escherichia coli (strain K12) plsC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7206-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017700

  • 3405.00€ / 1mg
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_417490.1
plsC Protein
Research Use Only
plsC
Recombinant Protein
E. coli
MLYIFRLIIT VIYSILVCVF GSIYCLFSPR NPKHVATFGH MFGRLAPLFG LKVECRKPTD AESYGNAIYI ANHQNNYDMV TASNIVQPPT VTVGKKSLLW IPFFGQLYWL TGNLLIDRNN RTKAHGTIAE VVNHFKKRRI SIWMFPEGTR SRGRGLLPFK TGAFHAAIAA GVPIIPVCVS TTSNKINLNR LHNGLVIVEM LPPIDVSQYG KDQVRELAAH CRSIMEQKIA ELDKEVAERE AAGKV
Greater than 90% as determined by SDS-PAGE.
947496
1 mg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers