missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Ornithine decarboxylase, inducible Protein
A DNA sequence encoding the Escherichia coli (strain K12) Ornithine decarboxylase, inducible, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8883-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_415220.1 | |
Ornithine decarboxylase, inducible Protein | |
Research Use Only | |
speF | |
Recombinant Protein | |
E. coli | |
MSKLKIAVSD SCPDCFTTQR ECIYINESRN IDVAAIVLSL NDVTCGKLDE IDATGYGIPV FIATENQERV PAEYLPRISG VFENCESRRE FYGRQLETAA SHYETQLRPP FFRALVDYVN QGNSAFDCPG HQGGEFFRRH PAGNQFVEYF GEALFRADLC NADVAMGDLL IHEGAPCIAQ QHAAKVFNAD KTYFVLNGTS SSNKVVLNAL LTPGDLVLFD RNNHKSNHHG ALLQAGATPV YLETARNPYG FIGGIDAHCF EESYLRELIA EVAPQRAKEA RPFRLAVIQL GTYDGTIYNA RQVVDKIGHL CDYILFDSAW VGYEQFIPMM ADCSPLLLDL NENDPGILVT QSVHKQQAGF SQTSQIHKKD SHIKGQQRYV PHKRMNNAFM MHASTSPFYP LFAALNINAK MHEGVSGRNM WMDCVVNGIN ARKLILDNCQ HIRPFVPELV DGKPWQSYET AQIAVDLRFF QFVPGEHWHS FEGYAENQYF VDPCKLLLTT PGIDARNGEY EAFGVPATIL ANFLRENGVV PEKCDLNSIL FLLTPAEDMA KLQQLVALLV RFEKLLESDA PLAEVLPSIY KQHEERYAGY TLRQLCQEMH DLYARHNVKQ LQKEMFRKEH FPRVSMNPQE ANYAYLRGEV ELVRLPDAEG RIAAEGALPY PPGVLCVVPG EIWGGAVLRY FSALEEGINL LPGFAPELQG VYIEEHDGRK QVWCYVIKPR DAQSTLLKGE KL | |
Greater than 90% as determined by SDS-PAGE. |
945297 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |