missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Ornithine carbamoyltransferase chain F Protein

A DNA sequence encoding the Escherichia coli (strain K12) Ornithine carbamoyltransferase chain F, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8748-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016271

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_414807.1
Ornithine carbamoyltransferase chain F Protein
Research Use Only
argF
Recombinant Protein
E. coli
SDLYKKHFLK LLDFTPAQFT SLLTLAAQLK ADKKNGKEVQ KLTGKNIALI FEKDSTRTRC SFEVAAFDQG ARVTYLGPSG SQIGHKESIK DTARVLGRMY DGIQYRGHGQ EVVETLAQYA GVPVWNGLTN EFHPTQLLAD LMTMQEHLPG KAFNEMTLVY AGDARNNMGN SMLEAAALTG LDLRLLAPKA CWPEESLVAE CSALAEKHGG KITLTEDVAA GVKGADFIYT DVWVSMGEAK EKWAERIALL RGYQVNAQMM ALTDNPNVKF LHCLPAFHDD QTTLGKQMAK EFDLHGGMEV TDEVFESAAS IVFDQAENRM HTIKAVMMAT LGE
Greater than 90% as determined by SDS-PAGE.
944844
10 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers