missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Mcr-1 Protein
A DNA sequence encoding the Escherichia coli (strain K12) Mcr-1, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | WP_049589868.1 |
---|---|
Name | Mcr-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | mcr1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16040551
|
enQuireBio™
QP7746-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16060551
|
enQuireBio™
QP7746-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16050551
|
enQuireBio™
QP7746-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16070551
|
enQuireBio™
QP7746-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16080551
|
enQuireBio™
QP7746-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16090551
|
enQuireBio™
QP7746-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
WP_049589868.1 | |
Research Use Only | |
mcr1 | |
Recombinant Protein | |
E. coli | |
MMQHTSVWYR RSVSPFVLVA SVAVFLTATA NLTFFDKISQ TYPIADNLGF VLTIAVVLFG AMLLITTLLS SYRYVLKPVL ILLLIMGAVT SYFTDTYGTV YDTTMLQNAL QTDQAETKDL LNAAFIMRII GLGVLPSLLV AFVKVDYPTW GKGLMRRLGL IVASLALILL PVVAFSSHYA SFFRVHKPLR SYVNPIMPIY SVGKLASIEY KKASAPKDTI YHAKDAVQAT KPDMRKPRLV VFVVGETARA DHVSFNGYER DTFPQLAKID GVTNFSNVTS CGTSTAYSVP CMFSYLGADE YDVDTAKYQE NVLDTLDRLG VSILWRDNNS DSKGVMDKLP KAQFADYKSA TNNAICNTNP YNECRDVGML VGLDDFVAAN NGKDMLIMLH QMGNHGPAYF KRYDEKFAKF TPVCEGNELA KCEHQSLINA YDNALLATDD FIAQSIQWLQ THSNAYDVSM LYVSDHGESL GENGVYLHGM PNAFAPKEQR SVPAFFWTDK QTGITPMATD TVLTHDAITP TLLKLFDVTA DKVKDRTAFI R | |
Greater than 90% as determined by SDS-PAGE. |
Mcr-1 Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |