missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Lipid kinase YegS Protein

A DNA sequence encoding the Escherichia coli (strain K12) Lipid kinase YegS, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6965-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16066900

  • 3745.00€ / 1mg
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_416590.1
Lipid kinase YegS Protein
Research Use Only
yegS
Recombinant Protein
E. coli
MAEFPASLLI LNGKSTDNLP LREAIMLLRE EGMTIHVRVT WEKGDAARYV EEARKFGVAT VIAGGGDGTI NEVSTALIQC EGDDIPALGI LPLGTANDFA TSVGIPEALD KALKLAIAGD AIAIDMAQVN KQTCFINMAT GGFGTRITTE TPEKLKAALG SVSYIIHGLM RMDTLQPDRC EIRGENFHWQ GDALVIGIGN GRQAGGGQQL CPNALINDGL LQLRIFTGDE ILPALVSTLK SDEDNPNIIE GASSWFDIQA PHDITFNLDG EPLSGQNFHI EILPAALRCR LPPDCPLLR
Greater than 90% as determined by SDS-PAGE.
946626
1 mg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers