missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Glutamate decarboxylase alpha Protein

A DNA sequence encoding the Escherichia coli (strain K12) Glutamate decarboxylase alpha, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6947-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16086820

  • 3745.00€ / 1mg
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_417974.1
Glutamate decarboxylase alpha Protein
Research Use Only
gadA
Recombinant Protein
E. coli
MDQKLLTDFR SELLDSRFGA KAISTIAESK RFPLHEMRDD VAFQIINDEL YLDGNARQNL ATFCQTWDDE NVHKLMDLSI NKNWIDKEEY PQSAAIDLRC VNMVADLWHA PAPKNGQAVG TNTIGSSEAC MLGGMAMKWR WRKRMEAAGK PTDKPNLVCG PVQICWHKFA RYWDVELREI PMRPGQLFMD PKRMIEACDE NTIGVVPTFG VTYTGNYEFP QPLHDALDKF QADTGIDIDM HIDAASGGFL APFVAPDIVW DFRLPRVKSI SASGHKFGLA PLGCGWVIWR DEEALPQELV FNVDYLGGQI GTFAINFSRP AGQVIAQYYE FLRLGREGYT KVQNASYQVA AYLADEIAKL GPYEFICTGR PDEGIPAVCF KLKDGEDPGY TLYDLSERLR LRGWQVPAFT LGGEATDIVV MRIMCRRGFE MDFAELLLED YKASLKYLSD HPKLQGIAQQ NSFKHT
Greater than 90% as determined by SDS-PAGE.
948027
1 mg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers