missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli D-cysteine desulfhydrase Protein
A DNA sequence encoding the Escherichia coli (strain K12) D-cysteine desulfhydrase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8917-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_416429.4 | |
D-cysteine desulfhydrase Protein | |
Research Use Only | |
dcyD | |
Recombinant Protein | |
E. coli | |
PLHNLTRFPR LEFIGAPTPL EYLPRFSDYL GREIFIKRDD VTPMAMGGNK LRKLEFLAAD ALREGADTLI TAGAIQSNHV RQTAAVAAKL GLHCVALLEN PIGTTAENYL TNGNRLLLDL FNTQIEMCDA LTDPNAQLEE LATRVEAQGF RPYVIPVGGS NALGALGYVE SALEIAQQCE GAVNISSVVV ASGSAGTHAG LAVGLEHLMP ESELIGVTVS RSVADQLPKV VNLQQAIAKE LELTASAEIL LWDDYFAPGY GVPNDEGMEA VKLLARLEGI LLDPVYTGKA MAGLIDGISQ KRFKDEGPIL FIHTGGAPAL FAYHPHV | |
Greater than 90% as determined by SDS-PAGE. |
946831 | |
10 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |