missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Aerobic glycerol-3-phosphate dehydrogenase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Aerobic glycerol-3-phosphate dehydrogenase, was expressed in the hosts and tags indicated.
352.00€ - 4075.00€
Specifications
Accession Number | NP_417884.1 |
---|---|
Gene ID (Entrez) | 947934 |
Name | Aerobic glycerol-3-phosphate dehydrogenase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16027190
|
enQuireBio™
QP7047-YE-100UG |
100 μg |
1100.00€
100µg |
Estimated Shipment:
Log in to see stock. |
|||||
16047190
|
enQuireBio™
QP7047-YE-1MG |
1 mg |
4075.00€
1mg |
Estimated Shipment:
Log in to see stock. |
|||||
16037190
|
enQuireBio™
QP7047-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16057190
|
enQuireBio™
QP7047-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16067190
|
enQuireBio™
QP7047-YE-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16077190
|
enQuireBio™
QP7047-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_417884.1 | |
Aerobic glycerol-3-phosphate dehydrogenase Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
947934 | |
Research Use Only | |
glpD | |
Recombinant Protein | |
Yeast | |
METKDLIVIG GGINGAGIAA DAAGRGLSVL MLEAQDLACA TSSASSKLIH GGLRYLEHYE FRLVSEALAE REVLLKMAPH IAFPMRFRLP HRPHLRPAWM IRIGLFMYDH LGKRTSLPGS TGLRFGANSV LKPEIKRGFE YSDCWVDDAR LVLANAQMVV RKGGEVLTRT RATSARRENG LWIVEAEDID TGKKYSWQAR GLVNATGPWV KQFFDDGMHL PSPYGIRLIK GSHIVVPRVH TQKQAYILQN EDKRIVFVIP WMDEFSIIGT TDVEYKGDPK AVKIEESEIN YLLNVYNTHF KKQLSRDDIV WTYSGVRPLC DDESDSPQAI TRDYTLDIHD ENGKAPLLSV FGGKLTTYRK LAEHALEKLT PYYQGIGPAW TKESVLPGGA IEGDRDDYAA RLRRRYPFLT ESLARHYART YGSNSELLLG NAGTVSDLGE DFGHEFYEAE LKYLVDHEWV RRADDALWRR TKQGMWLNAD QQSRVSQWLV EYTQQRLSLA S | |
Greater than 90% as determined by SDS-PAGE. |