missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 50S ribosomal protein L35 Protein

A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L35, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8626-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055651

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_416232.3
50S ribosomal protein L35 Protein
Research Use Only
rpmI
Recombinant Protein
E. coli
VRGAAKRFKK TGKGGFKHKH ANLRHILTKK ATKRKRHLRP KAMVSKGDLG LVIACLPY
Greater than 90% as determined by SDS-PAGE.
32367223
500 μg
Not determined.
Partial (see sequence information for more details).
E. coli
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers