missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 50S ribosomal protein L32 Protein

A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L32, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8630-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035671

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_415607.1
50S ribosomal protein L32 Protein
Research Use Only
rpmF
Recombinant Protein
E. coli
AVQQNKPTRS KRGMRRSHDA LTAVTSLSVD KTSGEKHLRH HITADGYYRG RKVIAK
Greater than 90% as determined by SDS-PAGE.
31659212
500 μg
Not determined.
Full Length
E. coli
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers