missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 30S ribosomal protein S21 Protein

A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S21, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8616-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015591

  • 985.00€ / 100µg
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_417537.1
30S ribosomal protein S21 Protein
Research Use Only
rpsU
Recombinant Protein
E. coli
PVIKVRENEP FDVALRRFKR SCEKAGVLAE VRRREFYEKP TTERKRAKAS AVKRHAKKLA RENARRTRLY
Greater than 90% as determined by SDS-PAGE.
947577
100 μg
Not determined.
Full Length
E. coli
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers