missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken Vesicle-associated membrane protein 1

A DNA sequence encoding the Gallus gallus (Chicken) Vesicle-associated membrane protein 7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6875-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016470

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001026292.1
Vesicle-associated membrane protein 1
Research Use Only
VAMP7
Recombinant Protein
E. coli
AILFAVVARG TTILAKHAWC GGNFLEVTEQ ILAKIPSENN KLTYSHGNYL FHYICQDRII YLCITDDDFE RSRAFNFLNE IKKRFQTTYG SRAQTALPYA MNSEFSSVLA AQLKYHSESK GTDQVAETQA QVDELKGIMV RNIDLVAQRG EKLELLIDKT ENLVDSSVTF KTTSRNLARA
Greater than 90% as determined by SDS-PAGE.
422297
500 μg
Not determined.
Cytoplasmic Domain
Chicken
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers