missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken Riboflavin-binding Protein

A DNA sequence encoding the Gallus gallus (Chicken) Riboflavin-binding protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8851-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026831

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_990794.1
Riboflavin-binding Protein
Research Use Only
RBP
Recombinant Protein
Yeast
QQYGCLEGDT HKANPSPEPN MHECTLYSES SCCYANFTEQ LAHSPIIKVS NSYWNRCGQL SKSCEDFTKK IECFYRCSPH AARWIDPRYT AAIQSVPLCQ SFCDDWYEAC KDDSICAHNW LTDWERDESG ENHCKSKCVP YSEMYANGTD MCQSMWGESF KVSESSCLCL QMNKKDMVAI KHLLSESSEE SSSMSSSEEH ACQKKLLK
Greater than 90% as determined by SDS-PAGE.
396449
10 μg
Not determined.
Partial (see sequence information for more details).
Chicken
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers