missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. trachomatis 60 kDa chaperonin Protein
A DNA sequence encoding the Chlamydia trachomatis (strain D/UW-3/Cx) 60 kDa chaperonin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7010-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_219613.1 | |
60 kDa chaperonin Protein | |
Research Use Only | |
groL | |
Recombinant Protein | |
E. coli | |
MVAKNIKYNE EARKKIQKGV KTLAEAVKVT LGPKGRHVVI DKSFGSPQVT KDGVTVAKEV ELADKHENMG AQMVKEVASK TADKAGDGTT TATVLAEAIY TEGLRNVTAG ANPMDLKRGI DKAVKVVVDQ IRKISKPVQH HKEIAQVATI SANNDAEIGN LIAEAMEKVG KNGSITVEEA KGFETVLDIV EGMNFNRGYL SSYFATNPET QECVLEDALV LIYDKKISGI KDFLPVLQQV AESGRPLLII AEDIEGEALA TLVVNRIRGG FRVCAVKAPG FGDRRKAMLE DIAILTGGQL ISEELGMKLE NANLAMLGKA KKVIVSKEDT TIVEGMGEKE ALEARCESIK KQIEDSSSDY DKEKLQERLA KLSGGVAVIR VGAATEIEMK EKKDRVDDAQ HATIAAVEEG ILPGGGTALI RCIPTLEAFL PMLTNEDEQI GARIVLKALS APLKQIAANA GKEGAIIFQQ VMSRSANEGY DALRDAYTDM LEAGILDPAK VTRSALESAA SVAGLLLTTE ALIAEIPEEK PAAAPAMPGA GMDY | |
Greater than 90% as determined by SDS-PAGE. |
884030 | |
50 μg | |
Not determined. | |
Full Length | |
C. trachomatis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |