missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. trachomatis 60 kDa chaperonin Protein

A DNA sequence encoding the Chlamydia trachomatis (strain D/UW-3/Cx) 60 kDa chaperonin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7010-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007030

  • 2360.00€ / 500µg
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_219613.1
60 kDa chaperonin Protein
Research Use Only
groL
Recombinant Protein
E. coli
MVAKNIKYNE EARKKIQKGV KTLAEAVKVT LGPKGRHVVI DKSFGSPQVT KDGVTVAKEV ELADKHENMG AQMVKEVASK TADKAGDGTT TATVLAEAIY TEGLRNVTAG ANPMDLKRGI DKAVKVVVDQ IRKISKPVQH HKEIAQVATI SANNDAEIGN LIAEAMEKVG KNGSITVEEA KGFETVLDIV EGMNFNRGYL SSYFATNPET QECVLEDALV LIYDKKISGI KDFLPVLQQV AESGRPLLII AEDIEGEALA TLVVNRIRGG FRVCAVKAPG FGDRRKAMLE DIAILTGGQL ISEELGMKLE NANLAMLGKA KKVIVSKEDT TIVEGMGEKE ALEARCESIK KQIEDSSSDY DKEKLQERLA KLSGGVAVIR VGAATEIEMK EKKDRVDDAQ HATIAAVEEG ILPGGGTALI RCIPTLEAFL PMLTNEDEQI GARIVLKALS APLKQIAANA GKEGAIIFQQ VMSRSANEGY DALRDAYTDM LEAGILDPAK VTRSALESAA SVAGLLLTTE ALIAEIPEEK PAAAPAMPGA GMDY
Greater than 90% as determined by SDS-PAGE.
884030
500 μg
Not determined.
Full Length
C. trachomatis
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers