missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. elegans ATP-dependent (S)-NAD(P)H-hydrate dehydratase Protein
A DNA sequence encoding the Caenorhabditis elegans ATP-dependent (S)-NAD(P)H-hydrate dehydratase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7162-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_499001.1 | |
ATP-dependent (S)-NAD(P)H-hydrate dehydratase Protein | |
Research Use Only | |
R107.2 | |
Recombinant Protein | |
E. coli | |
MDHFIKLLPK LTPHLRKGDC GKMGVIGGSL EYTGAPYFAA SSASRLGADL IHIFCDPDAA QVIKGYSPDL IVHPGMTANS IIPKLSRMDA IVIGPGLGRN PNIWPLMQEL FEFVRNRDVP FVIDGDGLWF VSEHIEKFPR QMSATVLTPN IVEFSRLCKS ALGEEDVLNV RNNSQLQHLA AELSRKMNVT IYLKGEVDLV VTPNGEVSKC STESSLRRCG GQGDVTAGSL GLFLYWAKKN LGDDWTSAHH EAGIASSWLV RTAGRRAFEK HGRSMNTPLL LDEIPKLVRD VETREMKDTV HTDSSKH | |
Greater than 90% as determined by SDS-PAGE. |
176277 | |
50 μg | |
Not determined. | |
Full Length | |
C. elegans | |
His-SUMO | |
Tris-based buffer, 50% glycerol |