missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. botulinum Flagellin Protein

A DNA sequence encoding the Clostridium botulinum (strain Hall / ATCC 3502 / NCTC 13319 / Type A) Flagellin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6926-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036750

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

WP_011987021.1
Flagellin Protein
Research Use Only
CBO2730
Recombinant Protein
E. coli
MIINHNLNAM NAHRQMAINT GNNGKAIEKL SSGLRINRAG DDAAGLAISE KMRGQIRGLN QASRNSQDAI SLIQTAEGAL NETHSILQRM RELTVQAAND TNVTVDRENL QKEVAELQSE INRISSQTQF NTKSLINGAV SKDADAMTFQ IGANSGQTIK LTIGSMDAKT LNVDKIKLDD TAANVTKQIA TIDNAINSVS KERAKLGANQ NRLEHTIANL DNSAENLQAA ESRIRDVDMA KEMMNFTKTN ILTQAAQAML AQANQAPQGV LQLLR
Greater than 90% as determined by SDS-PAGE.
5204320
500 μg
Not determined.
Full Length
C. botulinum
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers