missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Mannose-binding protein 1
A DNA sequence encoding the Bos taurus (Bovine) Mannose-binding protein C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9267-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_776532.1 | |
Mannose-binding protein 1 | |
Research Use Only | |
MBL | |
Recombinant Protein | |
Yeast | |
NCENIRKTCP VIACGPPGIN GIPGKDGRDG AKGEKGEPGQ GLRGSQGPPG KMGPQGTPGI PGIPGPIGQK GDPGENMGDY IRLATSERAT LQSELNQIKN WLIFSLGKRV GKKAFFTNGK KMPFNEVKTL CAQFQGRVAT PMNAEENRAL KDLVTEEAFL GITDQETEGK FVDLTGKGVT YQNWNDGEPN NASPGEHCVT LLSDGTWNDI ACSASFLTVC EFS | |
Greater than 90% as determined by SDS-PAGE. |
281297 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |