missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Lingual antimicrobial peptide Protein
A DNA sequence encoding the Bos taurus (Bovine) Lingual antimicrobial peptide, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7627-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_982259.3 | |
Lingual antimicrobial peptide Protein | |
Research Use Only | |
LAP | |
Recombinant Protein | |
E. coli | |
VRNSQSCRRN KGICVPIRCP GSMRQIGTCL GAQVKCCRRK | |
Greater than 90% as determined by SDS-PAGE. |
403090 | |
500 μg | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |