missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Interferon alpha-G Protein
A DNA sequence encoding the Bos taurus (Bovine) Interferon alpha-G, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7218-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001165511.1 | |
Interferon alpha-G Protein | |
Research Use Only | |
IFNAG | |
Recombinant Protein | |
Yeast | |
CHLPHTHSLA NRRVLTLLRQ LRRVSPSSCL QDRNDFAFPQ EALGGSQLQK AQAISVLHEV TQHTFQFFSV EGSAVVWDES LLDKLRDALD QQLTDLQFCL RQEEGLRGAP LLKEDSSLAV RKYFHRLTLY LQEKRHSPCA WEVVRAEVMR AFSSSTNLQE RFRRKD | |
Greater than 90% as determined by SDS-PAGE. |
100329206 | |
200 μg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |