missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Heterogeneous nuclear ribonucleoproteins A2 / B1 Protein
A DNA sequence encoding the Bos taurus (Bovine) Heterogeneous nuclear ribonucleoproteins A2/B1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7636-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001039440.1 | |
Heterogeneous nuclear ribonucleoproteins A2 / B1 Protein | |
Research Use Only | |
HNRNPA2B1 | |
Recombinant Protein | |
E. coli | |
MEREKEQFRK LFIGGLSFET TEESLRNYYE QWGKLTDCVV MRDPASKRSR GFGFVTFSSM AEVDAAMAAR PHSIDGRVVE PKRAVAREES GKPGAHVTVK KLFVGGIKED TEEHHLRDYF EEYGKIDTIE IITDRQSGKK RGFGFVTFDD HDPVDKIVLQ KYHTINGHNA EVRKALSRQE MQEVQSSRSG RGGNFGFGDS RGGGGNFGPG PGSNFRGGSD GYGSGRGFGD GYNGYGGGPG GGNFGGSPGY GGGRGGYGGG GPGYGNQGGG YGGGYDNYGG GNYGSGNYND FGNYNQQPSN YGPMKSGNFG GSRNMGGPYG GGNYGPGGSG GSGGYGGRSR Y | |
Greater than 90% as determined by SDS-PAGE. |
507564 | |
500 μg | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |