missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Cathelicidin-4 Protein

A DNA sequence encoding the Bos taurus (Bovine) Cathelicidin-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7211-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007730

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_777252.1
Cathelicidin-4 Protein
Research Use Only
CATHL4
Recombinant Protein
E. coli
ILPWKWPWWP WRR
Greater than 90% as determined by SDS-PAGE.
282166
10 μg
Not determined.
Full Length
Bovine
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers