missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacteriophage Shiga-like toxin 1 subunit B Protein

A DNA sequence encoding the Enterobacteria phage H19B (Bacteriophage H19B) Shiga-like toxin 1 subunit B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6941-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16086790

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

Shiga-like toxin 1 subunit B Protein
Research Use Only
stxB
Recombinant Protein
Yeast
TPDCVTGKVE YTKYNDDDTF TVKVGDKELF TNRWNLQSLL LSAQITGMTV TIKTNACHNG GGFSEVIFR
Greater than 90% as determined by SDS-PAGE.
10 μg
Not determined.
Full Length
Bacteriophage
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers