missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacillus Monomeric sarcosine oxidase Protein
A DNA sequence encoding the Bacillus sp. (strain NS-129) Monomeric sarcosine oxidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7152-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
Monomeric sarcosine oxidase Protein | |
Research Use Only | |
soxA | |
Recombinant Protein | |
E. coli | |
MSTHFDVIVV GAGSMGMAAG YYLAKQGVKT LLVDSFDPPH TNGSHHGDTR IIRHAYGEGR EYVPFALRAQ ELWYELEKET HHKIFTQTGV LVYGPKGGSA FVSETMEAAN IHSLEHELFE GKQLTDRWAG VEVPDNYEAI FEPNSGVLFS ENCIQAYREL AEAHGATVLT YTPVEDFEVT EDLVTIKTAK GSYTANKLVV SMGAWNSKLL SKLDVEIPLQ PYRQVVGFFE CDEAKYSNNA HYPAFMVEVE NGIYYGFPSF GGSGLKIGYH SYGQQIDPDT INREFGAYPE DEANLRKFLE QYMPGANGEL KKGAVCMYTK TPDEHFVIDL HPKYSNVAIA AGFSGHGFKF SSVVGETLAQ LATTGKTEHD ISIFSLNRDA LKKEAVK | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Bacillus | |
His | |
Tris-based buffer, 50% glycerol |