missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacillus Monomeric sarcosine oxidase Protein

A DNA sequence encoding the Bacillus sp. (strain NS-129) Monomeric sarcosine oxidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7152-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007500

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

Monomeric sarcosine oxidase Protein
Research Use Only
soxA
Recombinant Protein
E. coli
MSTHFDVIVV GAGSMGMAAG YYLAKQGVKT LLVDSFDPPH TNGSHHGDTR IIRHAYGEGR EYVPFALRAQ ELWYELEKET HHKIFTQTGV LVYGPKGGSA FVSETMEAAN IHSLEHELFE GKQLTDRWAG VEVPDNYEAI FEPNSGVLFS ENCIQAYREL AEAHGATVLT YTPVEDFEVT EDLVTIKTAK GSYTANKLVV SMGAWNSKLL SKLDVEIPLQ PYRQVVGFFE CDEAKYSNNA HYPAFMVEVE NGIYYGFPSF GGSGLKIGYH SYGQQIDPDT INREFGAYPE DEANLRKFLE QYMPGANGEL KKGAVCMYTK TPDEHFVIDL HPKYSNVAIA AGFSGHGFKF SSVVGETLAQ LATTGKTEHD ISIFSLNRDA LKKEAVK
Greater than 90% as determined by SDS-PAGE.
200 μg
Not determined.
Full Length
Bacillus
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers