Learn More
Julabo™ Recirculating Cooling Baths with Water Compressor
Product Code. p-8002694
Click to view available options
Flow Rate:
20 L/min.
22 L/min.
23 L/min.
28 L/min.
33 L/min.
40 L/min.
60 L/min.
80 L/min.
Capacity (Metric):
12 to 17 L
15 to 37 L
21 to 33 L
24 to 30 L
39 to 47 L
43 to 60 L
6 to 8 L
8 to 11 L
Unit Size:
Each
Product Code. 11944818
Brand: Julabo™ 9671017
Used for routine cooling applications both within laboratory and wider industrial workplace. Julabo™ Recirculating Cooling Baths with Water Compressor has large, bright LED display.
  • Ergonomic design and easy operation
  • Splashproof keypad with integrated mains switch
  • Reliable microprocessor PID temperature control
  • Filling level indicator
  • Powerful immersion pumps, suitable for continuous operation
  • Permissible temperature in return line +80°C
  • Easy filling from the top with hinged protective lid
  • Low liquid level protection with optical and audible alarm signal
  • Integrated stainless steel bath tanks
  • Removable venting grid for cleaning of the condenser
  • Front drain
  • No side vents
  • RS232 interface for PC-connection
  • IP class according to IEC 60529: 21
  • Pressure indicator
  • Adjustable bypass for pump pressure
  • Water cooled
  • Alarm output, potential-free change-over contact (max. 30 VA)
Specifications
Capacity (Metric) 12 to 17 L
Certifications/Compliance CE, IEC 60529: 21
Cooling Capacity 0.4kW at -20°C, 0.85kW at -10°C, 1.1kW at 0°C, 1.5kW at 10°C, 1.7kW at 20°C (Med
Flow Rate 23 L/min.
For Use With (Equipment) For environmentally friendly cooling
Temperature Range 5°C to 40°C
Type Recirculating Cooling Bath
Max. Pressure 14.5 psi
Model FLW1701
Refrigerant R404A
Temperature Stability ±0.5°C
Length (Metric) 76 cm
Width (Metric) 50 cm
Height (Metric) 64 cm
Weight (Metric) 85 kg
Electrical Requirements 115/230 V 50/60 Hz
Voltage 115/230 V
Frequency 50/60 Hz