Learn More
Lenz Laborglasintrumente™ Receiving Flasks with PUR Coating
Product Code. 11963391
Change view
Click to view available options
Volume (Metric):
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11963391 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11963391 Supplier Lenz Laborglasintrumente™ Supplier No. 03064037
  • ONLINE EXCLUSIVE PRICE
    127.00€
    87.95€ / Pack of 1
    Save 39.05€
    30% Off
Estimated Shipment: 04-01-2027
Receiving flasks especially for rotary evaporators
  • With spherical joint socket S 35
  • With highly transparent PUR coating
  • Autoclavable
  • Material: DURAN™
Specifications
Connection Type Socket 35
For Use With (Equipment) Rotary Evaporators
Quantity 1
Volume (Metric) 100 mL
Diameter (Metric) 64 mm