Learn More
Lenz Laborglasinstrumente™ Receiving Flask
Product Code. 11806114
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
2000 mL
Unit Size:
Each
Product Code. 11806114
Brand: Lenz Laborglasinstrumente™ 03063549
Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ Receiving Flasks are manufactured from durable DURAN™ glass.
  • With spherical joint socket S 35
  • With inscription label
  • Available in a range of capacities
Specifications
Material Duran Glass
Type Receiving Flask
For Use With (Equipment) Rotary Evaporators
Volume (Metric) 250 mL
Diameter (Metric) 85 mm