Learn More
Macherey-Nagel™ Receiver Plate
Use for filtration in 96-well format. Macherey Nagel™ Receiver Plates are important accessories for high throughput work.
Brand: Macherey-Nagel™ 740686.4
Additional Details : Weight : 0.56700kg
Description
- Receiver plate accessory
- High-throughput applications
- Filter frits
- Suitable for centrifugation and vacuum
Specifications
96-well Plate | |
Inserted filter frits of 20μm pore size |