Learn More
Macherey-Nagel™ Receiver Plate
Use for filtration in 96-well format. Macherey Nagel™ Receiver Plates are important accessories for high throughput work.
152.00€ - 186.00€
Specifications
Format 96-well Plate
View More Specs
Products 4
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
11942542
View Documents Promotion Details
Macherey-Nagel™
740688.4
Inserted filter frits of 50μm pore size
183.00€
Pack of 4
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
11912542
View Documents Promotion Details
Macherey-Nagel™
740689.4
Inserted hydrophilized filter frits of 50μm pore size
186.00€
Pack of 4
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
11932542
View Documents Promotion Details
Macherey-Nagel™
740686.4
Inserted filter frits of 20μm pore size This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
11902542
View Documents Promotion Details
Macherey-Nagel™
740687.4
Inserted hydrophilized filter frits of 20μm pore size This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • Receiver plate accessory
  • High-throughput applications
  • Filter frits
  • Suitable for centrifugation and vacuum
Specifications
Specifications
96-well Plate