Learn More
Cytiva Ready-To-Go™ GenomiPhi HY DNA Amplification Kit
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60++g yield from nanogram amounts of DNA sample.
551.00€ - 1649.00€
Specifications
Content And Storage | Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons |
---|---|
Description | illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample. |
For Use With (Application) | Genomic DNA Amplification |
Size | > 10 kb |
Test Time | 4 h |
Product Code | Brand | Includes | No. of Reactions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | No. of Reactions | Price | Quantity & Availability | ||||
12987160
|
Cytiva
25-6603-24 |
1 x 2.6mL 2X Denaturation Buffer, 3 x 8-well strips Ready-To-Go™ GenomiPhi HY cakes, 1 x vial control DNA, 12 strips of 8 domed caps | 24 |
551.00€
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
12997160
|
Cytiva
25-6603-96 |
1 x 2.6mL 2X Denaturation Buffer, 3 x 96-well strips Ready-To-Go™ GenomiPhi HY cakes, 1 x vial control DNA, 12 strips of 8 domed caps | 96 |
1649.00€
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
||||
Description
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit is part of the Phi29 DNA polymerase family of products from Cytiva.
- Representative amplification of the whole genome.
- Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
- Quick and simple automation-friendly protocol; no thermal cycler required.
- Shipping and storage at room temperature provides easy access and reduced carbon footprint.
- Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).
Specifications
Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons | |
Genomic DNA Amplification | |
4 h |
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample. | |
> 10 kb |