Learn More
DiscoverX Ready-to-Assay™ XCR1 Chemokine Receptor Frozen Cells
Click to view available options
Quantity:
2 Vials of Cells
Unit Size:
2 ampoules
Description
The Ready-to-Assay™ XCR1 Chemokine Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. lymphotactin) based activation of XCR1 GPCR activity via detection of increasing levels of intracellular calcium. The included cells overexpress XCR1 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of XCR1 , the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the cells in a convenient thaw-and-use format.
Specifications
Specifications
| Assay Entry | XCR1 |
| For Use With (Application) | Frozen Cells |
| Primary Agonist | lymphotactin |
| Quantity | 2 Vials of Cells |
| Target Entry | XCR1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.