Learn More
DiscoverX Ready-to-Assay™ SST5 Somatostatin Receptor Frozen Cells
Product Code. 17325279
Encompass
Change view
Click to view available options
Quantity:
2 Vials of Cells
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17325279 2 Vials of Cells 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17325279 Supplier DiscoverX Supplier No. HTS139RTA
The Ready-to-Assay™ SST5 Somatostatin Receptor Frozen Cells measure SSTR5 (GPCR) activation via an increase in intracellular calcium. Ready-to-Assay kits contain two vials of thaw-and-use cells.
The Ready-to-Assay™ SST5 Somatostatin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. somatostatin) based activation of SSTR5 GPCR activity via detection of increasing levels of intracellular calcium. The included cells overexpress SSTR5 alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of SSTR5 , the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the cells in a convenient thaw-and-use format.
Specifications
Assay Entry SST5
For Use With (Application) Frozen Cells
Primary Agonist somatostatin
Quantity 2 Vials of Cells
Target Entry SSTR5