missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX Ready-to-Assay™ OX1 Orexin Receptor Frozen Cells

Product Code. 17385319
Encompass
Change view
Click to view available options
Quantity:
2 Vials of Cells
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17385319 2 Vials of Cells 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17385319 Supplier DiscoverX Supplier No. HTS175RTA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Ready-to-Assay™ OX1 Orexin Receptor Frozen Cells measure HCRTR1 (GPCR) activation via an increase in intracellular calcium. Ready-to-Assay kits contain two vials of thaw-and-use cells.

The Ready-to-Assay™ OX1 Orexin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. orexins) based activation of HCRTR1 GPCR activity via detection of increasing levels of intracellular calcium. The included cells overexpress HCRTR1 alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of HCRTR1 , the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the cells in a convenient thaw-and-use format.

Specifications

Assay Entry OX1
For Use With (Application) Frozen Cells
Primary Agonist orexins
Quantity 2 Vials of Cells
Target Entry HCRTR1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.