missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX Ready-to-Assay™ FP Prostanoid Receptor Frozen Cells
Description
The Ready-to-Assay™ FP Prostanoid Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. prostaglandin F2a) based activation of PTGFR GPCR activity via detection of increasing levels of intracellular calcium. The included cells overexpress PTGFR alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of PTGFR , the resulting calcium activity can be assayed in either detection mode. Ready-to-assay kits consist of two vials of the cells in a convenient thaw-and-use format.
Specifications
Specifications
| Assay Entry | FP |
| For Use With (Application) | Frozen Cells |
| Primary Agonist | prostaglandin F2a |
| Quantity | 2 Vials of Cells |
| Target Entry | PTGFR |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?