missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX Ready-to-Assay™ FFA2/GPR43 Free Fatty Acid Receptor Frozen Cells

Product Code. 17355099
Encompass
Click to view available options
Quantity:
2 Vials of Cells
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17355099

Brand: DiscoverX HTS063RTA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Ready-to-Assay™ FFA2/GPR43 Free Fatty Acid Receptor Frozen Cells measure FFAR2 (GPCR) activation via an increase in intracellular calcium. Ready-to-Assay kits contain two vials of thaw-and-use cells.

The Ready-to-Assay™ FFA2/GPR43 Free Fatty Acid Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. free fatty acids) based activation of FFAR2 GPCR activity via detection of increasing levels of intracellular calcium. The included cells overexpress FFAR2 alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of FFAR2 , the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the cells in a convenient thaw-and-use format.

Specifications

Assay Entry FFA2/GPR43
For Use With (Application) Frozen Cells
Primary Agonist free fatty acids
Quantity 2 Vials of Cells
Target Entry FFAR2
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.