Learn More
Afora™ Reactor Cover
Click to view available options
Diameter (Metric) Outer:
100 mm
Standard Taper Joint:
Center: 29/32, Sidearms: 14/24, 14/23, 14/24, 29/32
Center: 29/32, Sidearms: 14/24, 14/23, 29/32, 29/32
Unit Size:
Each
Description
- Reactor accessory
Specifications
Specifications
| Material | Classic Frosted Glass |
| Includes | Center arm: 29/32; Side arms: 14/24 (0°), 14/23 (5°), 14/24 (10°), 29/32 (15°) |
| Product Type | Reactor Cover |
| Standard Taper Joint | Center: 29/32, Sidearms: 14/24, 14/23, 14/24, 29/32 |
| Diameter (Metric) Outer | 100 mm |