Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical
Product Code. 11512204
Click to view available options
Capacity (Metric):
0.25 L
1 L
6 L
10 L
20 L
Diameter (Metric) Outer:
85 mm
131 mm
236 mm
280 mm
350 mm
Includes:
Lab Flange 100 LF
Lab Flange 150 LF
Lab Flange 200 LF
Lab Flange 60 LF
Unit Size:
Each
Product Code. 11512204
Brand: Lenz Laborglasintrumente 06005488
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Without graduation
  • With laboratory flange LF
  • With groove
  • Made of DURAN™ tubing
  • Other sizes on request
Specifications
Capacity (Metric) 10 L
Material Duran Glass
Height (Metric) 335 mm
Includes Lab Flange 150 LF
No. per Pack 1 Ea.
Diameter (Metric) Outer 280 mm